Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

EP1300

Sigma-Aldrich

Sigma-Aldrich® CVS10DSYS omniPAGE mini dual vertical electrophoresis system

10x10 cm, with caster

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000
NACRES:
NC.01

grade

electrophoresis grade

W × D × H

19 cm × 13 cm × 15 cm

volume × volume

min 250 mL × max 1200 mL

General description

The preferred unit for routine mini protein electrophoresis, the omniPAGE mini is constructed using the latest injection moulding manufacturing techniques. This gives a high quality, low cost product with unsurpassed finish,durability and strength. The unit incorporates a sealing system which is compatible with all major types of 8 x 10 cm and 10 x 10 cm precast gel. Gel casting and running utilize the same insert, no transfer of glass plates during gel casting is necessary. The insert contains the option of sliding clamps or just one screw per clamping bar side allowing very rapid set up of both handcast and precast gels. Ultra soft silicone seals and pressure bars which surround the glass plates guarantee leak proof gelcasting. 2 mm thick glass plates prevent breakage and have bonded spacers for convenience. Accessory electroblotting and tube gel modules are available which use the same outer tank and lid.

Included with the omniPAGE mini unit are dual 10 x 10 cm glass plates with bonded 1 mm thick spacers, 2 x 12 sample combs, cooling pack, dummy plate and casting base.

  • Typical Applications include Mini SDS-PAGE, Native PAGE, Gradient, Second dimension and Nucleic Acid Separation
  • Injection moulded construction
  • Compatible with all 8 x 10 and 10 x 10 cm precast gels
  • Rapid gel casting and loading
  • Low buffer volumes
  • Rapid set up cooling
  • Run up to four gels

Furthermore, these units can accommodate gels with the following dimensions (W x L): 7.5 x 8.5 cm. In addition, this unit accommodates a variety of comb options that include sample options of 1, 5, 8MC, 9, 10, 12, 16MC, and 20 as well as comb featuring 0.75, 1, 1.5 and 2.0 mm thickness.

Legal Information

SIGMA-ALDRICH is a registered trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service