Key Documents
EP1211
Sigma-Aldrich
Sigma-Aldrich® MSCHOICETRIO buffer saver blocks
2 blocks per package
About This Item
UNSPSC Code:
41100000
NACRES:
NC.01
Application
Physically reduce the volume of the gel chamber by up to 100 mL, each block displaces 50 mL buffer.
Legal Information
SIGMA-ALDRICH is a registered trademark of Merck KGaA, Darmstadt, Germany
Choose from one of the most recent versions:
Certificates of Analysis (COA)