Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EP1136

Sigma-Aldrich

Sigma-Aldrich® MSMINIDUO comb

16 sample, thickness 1.5 mm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000
NACRES:
NC.01

thickness

1.5 mm

well volume

15 μL

Looking for similar products? Visit Product Comparison Guide

Application

Available in 4 thicknesses and color coded:
Black combs: 0.75 mm for ultra-resolved bands
White combs: 1.0 mm supplied as a standard comb
Red combs: 1.5 mm for maximizing sample volume
Blue combs: 2.0 mm for maximizing sample volume

Analysis Note

Comb Prep number indicates the number of preparatory wells. Marker number indicates the number of small volume wells intested for molecular weight markers. MC indicates multichannel pipette compatible. All sample well volumes are based on a 5 mm thick agarose gel.

Legal Information

SIGMA-ALDRICH is a registered trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service