Key Documents
EP022822217
Rotor for Eppendorf® Vacufuge® Plus Concentrator
F-45-12-31, non aerosol tight, (12 x 20.0 mL flat-bottom tubes)
About This Item
UNSPSC Code:
41103909
feature
to hold 12 x 20 mL flat-bottom tubes
to hold 12 x 31 - 55 mm tubes
fixed angle rotor
non aerosol tight
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 022822217 (NA)
technique(s)
centrifugation: suitable
capacity
(12 x 20.0 mL flat-bottom tubes)
compatibility
for use with centrifuge models Vacufuge Plus Concentrator
General description
Selection of rotors for the Eppendorf Vacufuge Plus.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Vacufuge is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)