Key Documents
EP022638963
Eppendorf® CombiSlide Adapter
holds 12 slides, for plate rotors, pack of 2 ea
Select a Size
2 EA
$372.00
$372.00
Check Cart for Availability
Select a Size
Change View
2 EA
$372.00
About This Item
UNSPSC Code:
41103911
$372.00
Check Cart for Availability
feature
holds 12 slides, for plate rotors
packaging
pack of 2 ea
manufacturer/tradename
Eppendorf® 022638963
technique(s)
centrifugation: suitable
compatibility
for use with A-2-DWP
for use with A-4-62-MTP
for use with Rotors A-4-81 MTP/Flex
Related Categories
General description
This Eppendorf adapter is for use with the Eppendorf swing-bucket rotor for centrifuges 5804 / 5804 R. Fits 12 glass slides per adapter; 48 slides per rotor. Adapter is autoclavable (20 min, 121°C).
Features and Benefits
  • Safe centrifugation support for glass slides and microarrays for MTP and DWP rotors
  • Maximum of 12 slides per bucket
  • Also fits in A-2-DWP for Models 5804 and 5804 R
  • Most flexible: novel slot frames allow centrifugation either across or parallel to the rotational direction
  • Maximum rcf at 1,000 x g (for A-4-81-MTP/Flex and A-4-62-MTP)
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)