Key Documents
EP022492250
Eppendorf® epT.I.P.S.® Racks
sterile, volume × L 0.1-20 μL × 40 mm, gray, pack of 480 ea (5 racks of 96 tips)
About This Item
UNSPSC Code:
41121600
material
gray
sterility
sterile
feature
DNase free
RNase free
packaging
pack of 480 ea (5 racks of 96 tips)
manufacturer/tradename
Eppendorf® 022492250 (NA)
maximum volume
20 μL
volume × L
0.1-20 μL × 40 mm
foreign activity
PCR inhibitors, human and baterial DNA, DNase, RNase, ATP, Pyrogen, free
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
epT.I.P.S. is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)