Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EP0030742044

Eppendorf® Cell Imaging Slides

Tissue Culture (TC)-treated surface, wells 1, pack of 16 plates (individually wrapped)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122600
Product EP0030742044 is not currently sold in your country. Contact Technical Service

material

colorless
soda-lime glass

sterility

sterile

packaging

pack of 16 plates (individually wrapped)

manufacturer/tradename

Eppendorf® 0030742044

W × D × H

127.8 mm × 76.0 mm × 12.4 mm

wells

1

working volume

2.4-3.0 mL

binding type

Tissue Culture (TC)-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

The easy-to-open, securely closing, Flex Tubes offers comfort and dependability for sample preparation, centrifugation, and storage. One vessel for all applications in molecular biology, chemistry, and biochemistry.
Safer than ever: Flex Tubes can be centrifuged up to 30,000 × g without any problems!

Best material!
Eppendorf consumables incl. the Eppendorf Tubes 3810X are made exclusively from high quality, ultra pure polypropylene, which complies with FDA guidelines 21 CFR § 177.1520 »Olefin Polymers«, and 21 CFR § 178.2010 »Antioxidants and Stabilizers for Polymers«. Extensive quality checks are in place prior to and during production. Furthermore, the final product is examined for functionality. The functionality tests for tubes include lid closure security in a boiling water bath, centrifugation stability and vapor tightness. These quality controls and tests ensure the sustained high product quality across the entire production phase – lot by lot. Please see APPLICATION NOTE No.056 for details: The best material for original Eppendorf Tubes and Plates: Properties and chemical resistance of polypropyleneFlex Tubes

Highest sample integrity!
Manufactured without the use of slip agents, plasticizers, and biocides – substances that have been shown to leach from plastic consumables into the sample and negatively affect bio–assay results or measurements, e.g. nucleic acid quantification by photometric measurements at 260 nm.

Features and Benefits

Tube lid is easy to open and close for ergonomic operation

Polypropylene provides high resistance to chemicals, mechanical stress, and temperature extremes

g-Safe: Exceptional centrifugation stability up to 30,000 × g prevents sample loss due to tube breakage and provides extra safety when working with hazardous samples

Manufactured without the use of slip agents, plasticizers, and biocides – substances that have been shown to leach from plastic consumables into the sample and negatively affect bio-assay results

Graduation for simple verification of the pipetted volume

Frosted lid and surface on the side for easy labeling

Guaranteed functionality from -86 deg. C to 100 deg. C

Autoclavable when open (121 deg.C, 20 min)

Available in Eppendorf Quality and lot-certified PCR clean purity grade

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service