EP0030742010
Eppendorf® Cell Imaging Coverglasses
Tissue Culture (TC)-treated surface, wells 2, pack of 16 plates (individually wrapped)
All Photos(1)
About This Item
UNSPSC Code:
41122600
Recommended Products
material
borosilicate glass Type 1
colorless
sterility
sterile
packaging
pack of 16 plates (individually wrapped)
manufacturer/tradename
Eppendorf® 0030742010
W × D × H
127.8 mm × 76.0 mm × 12.4 mm
wells
2
working volume
1.3-1.8 mL
binding type
Tissue Culture (TC)-treated surface
General description
Eppendorf Cell Imaging Slides and Coverglasses offer a high chemical resistance of materials for reliable and reproducible results in fixation protocols. Tool-free and easy removal of chambers from Cell Imaging Slides and Coverglasses makes working easy, convenient and reliable. Products are available with one to eight chambers to suit a variety of applications.
Our rigorous quality assurance for Eppendorf Cell Imaging Consumables includes
› Proof on non-cytotoxicity according to ISO 10993 specifications
› Lot-certified sterility according to ISO 11137 and 11135
› Sterility assurance level (SAL) of 10-6
› Lot-specific testing and certification for absence of detectable pyrogens, DNA, DNase and RNase
› Testing for cell attachment and cell growth with an anchorage-dependent cell line
› Sterile, pyrogen-, DNase-, RNase-, human and bacterial DNA-free. Non-cytotoxic
Our rigorous quality assurance for Eppendorf Cell Imaging Consumables includes
› Proof on non-cytotoxicity according to ISO 10993 specifications
› Lot-certified sterility according to ISO 11137 and 11135
› Sterility assurance level (SAL) of 10-6
› Lot-specific testing and certification for absence of detectable pyrogens, DNA, DNase and RNase
› Testing for cell attachment and cell growth with an anchorage-dependent cell line
› Sterile, pyrogen-, DNase-, RNase-, human and bacterial DNA-free. Non-cytotoxic
Features and Benefits
Choose between a microscopic slide with 1 mm thickness or a 170 um coverglass, both providing excellent signal-to-noise ratios in fluorescent staining protocols
Facilitated growth of most adherent cell types due to innovative TC treated glass surface
Chambers and lids show high chemical resistance, so fixation even with acetone is possible without problems
Benefit from easy and tool-free removal of the chambers with minimized glue residues
Precise planarity supports high resolution microscopes and autofocus systems
Facilitated growth of most adherent cell types due to innovative TC treated glass surface
Chambers and lids show high chemical resistance, so fixation even with acetone is possible without problems
Benefit from easy and tool-free removal of the chambers with minimized glue residues
Precise planarity supports high resolution microscopes and autofocus systems
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)