Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EP0030741005

Eppendorf® Cell Imaging Plates

size 24 wells, bottom, 25 μm (film), pack of 20 plates (individually wrapped)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122107
Product EP0030741005 is not currently sold in your country. Contact Technical Service

material

black polystyrene
flat bottom clear wells

sterility

sterile

feature

bottom 25 μm (film)
lid
skirt (F-bottom with low skirt design)

packaging

pack of 20 plates (individually wrapped)

manufacturer/tradename

Eppendorf® 0030741005

parameter

300 × g max. RCF

technique(s)

PCR: suitable

size

24 wells

well volume

1.88 mL

working volume

0.5-1.0 mL

Looking for similar products? Visit Product Comparison Guide

General description

  • Eppendorf® Cell Imaging Plates of size 24 wells, bottom, capacity 25 μm(film), sterile purified, are Eppendorf cell imaging consumables with a low skirt design perfectly suited to access all wells with immersion objectives.
  • They have excellent planarity, which ensures unsurpassed manual handling and reliable results in automated devices.
  • They are also individually wrapped for reliable purity.
  • The ultrathin film bottom allows for high gas permeability and UV-light transparency, ideal for phototoxicity and hypoxic studies.
  • It also comprises of TC treated surfaces for efficient growth of adherent cells.

Eppendorf Cell Imaging Slides and Coverglasses offer a high chemical resistance of materials for reliable and reproducible results in fixation protocols. Tool-free and easy removal of chambers from Cell Imaging Slides and Coverglasses makes working easy, convenient and reliable. Products are available with one to eight chambers to suit a variety of applications.

Our rigorous quality assurance for Eppendorf Cell Imaging Consumables includes

› Proof on non-cytotoxicity according to ISO 10993 specifications
› Lot-certified sterility according to ISO 11137 and 11135
› Sterility assurance level (SAL) of 10-6
› Lot-specific testing and certification for absence of detectable pyrogens, DNA, DNase and RNase
› Testing for cell attachment and cell growth with an anchorage-dependent cell line
› Sterile, pyrogen-, DNase-, RNase-, human and bacterial DNA-free. Non-cytotoxic

Features and Benefits

Choose between a microscopic slide with 1 mm thickness or a 170 um coverglass, both providing excellent signal-to-noise ratios in fluorescent staining protocols

Facilitated growth of most adherent cell types due to innovative TC treated glass surface

Chambers and lids show high chemical resistance, so fixation even with acetone is possible without problems

Benefit from easy and tool-free removal of the chambers with minimized glue residues

Precise planarity supports high resolution microscopes and autofocus systems

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service