Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EP0030740009

Eppendorf® Cell Imaging Dish

sterile

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121812
Product EP0030740009 is not currently sold in your country. Contact Technical Service

material

glass
polystyrene

sterility

sterile

packaging

pkg of 30 pieces (2 dishes per bag)

manufacturer/tradename

Eppendorf® 0030740009

surface area

8.87 cm2

thickness

145 μm

working volume

0.5-0.75 mL

Looking for similar products? Visit Product Comparison Guide

General description

Eppendorf Cell Imaging Plates with film bottom have excellent light transmission rates even for UV-A and UV-B light. The autofluorescence of the material is lower than a conventional polystyrene bottom with a significant reduction in background signaling. Furthermore, the film bottom enables high gas transfer: oxygen supply and equilibration with the atmosphere are achieved directly through the plate bottom. The glass bottom plates offer excellent planarity for sophisticated microscopic analysis.

Our rigorous quality assurance for Eppendorf Cell Imaging Consumables includes

  • Proof on non-cytotoxicity according to ISO 10993 specifications
  • Lot-certified sterility according to ISO 11137 and 11135
  • Sterility assurance level (SAL) of 10-6
  • Lot-specific testing and certification for absence of detectable pyrogens, DNA, DNase and RNase
  • Testing for cell attachment and cell growth with an anchorage-dependent cell line
  • Sterile, pyrogen-, DNase-, RNase-, human and bacterial DNA-free. Non-cytotoxic

Features and Benefits

TC treated coverglass surface for reliable results when working with adherent cells

Easy access to the complete imaging area with immersion lenses for alleviated handling and advanced compatibility with microscopes

An 18 mm central cavity for defined growth and staining of cells for reduced costs of antibodies and dyes

Polygonal gripping zone for improved handling

Macroscopic orientation marks facilitate easy orientation/microinjection

Low rim side walls allow for easy access for microinjection
Available thickness: 145 um and 170 um

Easy access to the complete imaging area with immersion lenses for alleviated handling and advanced compatibility with microscopes

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service