EP0030722116
Eppendorf® Cell Culture Plates
size 24 wells, flat bottom clear wells, pkg of 60 plates (individually wrapped), sterile, with lid
About This Item
Recommended Products
material
flat bottom clear wells
polystyrene
sterility
sterile
feature
with lid
packaging
pkg of 60 plates (individually wrapped)
manufacturer/tradename
Eppendorf® 0030722116
technique(s)
PCR: suitable
W × D × H
127.8 mm × 85.5 mm × 20.0 mm
size
24 wells
surface area
20.89 cm2
well diam.
16.2 mm
well volume
3.6 mL
working volume
0.5-1 mL
Looking for similar products? Visit Product Comparison Guide
General description
The central 18 mm cavity is lower than the level of the surrounding polystyrene dish, which enables the concentration of the cells during seeding on the glass surface and helps to reduce costs of antibodies and dyes. Plate handling and reference are easy, safe, and convenient due to a polygonal gripping zone and directional orientation marking.
Rigorous quality assurance for Eppendorf Cell Imaging Consumables includes
- Proof on non-cytotoxicity according to ISO 10993 specifications
- Lot-certified sterility according to ISO 11137 and 11135
- Sterility assurance level (SAL) of 10-6
- Lot-specific testing and certification for absence of detectable pyrogens, DNA, DNase and RNase
- Testing for cell attachment and cell growth with an anchorage-dependent cell line
- Sterile, pyrogen-, DNase-, RNase-, human and bacterial DNA-free. Non-cytotoxic
Features and Benefits
Moat surrounding the outer wells to prevent the “edge effect” when filled with liquid
Chimney-well design enables filling of the inter-well spaces of the plate to level out well-to-well temperature shifts outside the incubator
Robust stacking performance by pronounced rims on plate lid and excellent fitting of lid and base when used in stacks
Lid taps under the plate for minimized surface contact and therefore reduced risk of contamination
Easy differentiation of lid and plate bottom due to a pronounced corrugation and wider sized base
Pronounced ventilation gaps for optimized gas and temperature transfer when incubating in stacks
Optimized microscopical performance due to enhanced planarity, reduction of meniscus and clarity of the material
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service