Key Documents
EP0030610334
Eppendorf® Lobind Microplate 384/V-PP, 384 wells
def. color (wells colorless), with barcode, polypropylene, pkg of 80 ea
About This Item
UNSPSC Code:
41122107
material:
polypropylene
size:
384 wells
Product EP0030610334 is not currently sold in your country. Contact Technical Service
material
polypropylene
packaging
pkg of 80 ea
manufacturer/tradename
Eppendorf® 0030610334
technique(s)
PCR: suitable
size
384 wells
General description
Maximize your sample recovery with Eppendorf LoBind Plates. Eppendorf LoBind Plates are designed with a special two-component polymer mix that creates a hydrophilic surface to ensure optimal sample recovery rates. These plates are specifically designed for research or sensitive testing methods where they enable excellent results due to less sample binding to the plate surface.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)