EP0030609700
Eppendorf® Microplate 96/F
black wells, PCR clean, white border
About This Item
material
polypropylene
sterility
sterile
feature
PCR clean
packaging
pkg of 80 plates (5 bags x 16 plates - barcoded)
manufacturer/tradename
Eppendorf® 0030609700
technique(s)
PCR: suitable
size
96 well
color
black wells
white border
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service