Key Documents
EP0030609115
Eppendorf® Microplate 96/F-PP, 96 wells
with barcode, def. color (wells colorless), PCR clean, polypropylene, sterile, pkg of 80 ea
About This Item
UNSPSC Code:
41122107
material:
polypropylene
size:
96 wells
sterility:
sterile
Product EP0030609115 is not currently sold in your country. Contact Technical Service
material
polypropylene
sterility
sterile
packaging
pkg of 80 ea
manufacturer/tradename
Eppendorf® 0030609115
technique(s)
PCR: suitable
size
96 wells
General description
Pipette, control and recover your samples with ease using Eppendorf Microplates! Designed with the easy-to-read alphanumeric OptiTrack matrix label to identify individual wells, speed up the filling process and significantly reduce the risk of pipetting errors. Eppendorf Microplates are made with high-transparency polypropylene ensuring quick and easy location of samples
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)