Key Documents
EP0030510100
Eppendorf® Deepwell plates, standard, 384 wells
capacity 200 μL, pkg of 120 ea
About This Item
UNSPSC Code:
41122107
size:
384 wells
Product EP0030510100 is not currently sold in your country. Contact Technical Service
packaging
pkg of 120 ea
manufacturer/tradename
Eppendorf® 0030510100
technique(s)
PCR: suitable
capacity
200 μL
size
384 wells
Features and Benefits
  • OptiTrack matrix: up to 30 % faster sample identification and fewer pipetting errors via high-contrast alphanumeric labeling
  • RecoverMax well design: Optimized well geometry for maximum sample recovery and excellent mixing properties
  • g-Safe: exceptional centrifugation stability up to 6,000 × g for faster protocols and improved sample quality
  • Raised well rims and smooth surface for reliable closing, even with heat sealing
  • Easy and reliable stacking
  • Minimized remaining volume and high uniformity from well to well
  • High-quality polypropylene ensures high resistance to chemicals, mechanical stress and temperature extremes
  • Manufactured without slip agents, plasticizers or biocides – Materials which could have a negative effect on bioassays
  • Autoclavable (121 C, 20 min)
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)