EP0030509218
Eppendorf® Deepwell plates, 96 wells
def. Color (wells colorless), with barcode, capacity 1000 μL, sterile, pkg of 80 ea
About This Item
Recommended Products
sterility
sterile
packaging
pkg of 80 ea
manufacturer/tradename
Eppendorf® 0030509218
technique(s)
PCR: suitable
capacity
1000 μL
size
96 wells
Looking for similar products? Visit Product Comparison Guide
General description
- Eppendorf® Deepwell plates, 96 wells with 1000 μL capacity are available as sterile and are used for sample preparation/storage applications.
- They are manufactured from high-quality polypropylene which guarantees high resistance to chemicals, mechanical stress and temperature extremes.
- OptiTrack®matrix: up to 30% faster sample identification and fewer pipetting errors via high-contrast alphanumeric labeling.
- RecoverMax® well design: Optimized well geometry for maximum sample recovery and excellent mixing properties.
- Minimal remaining volume and high uniformity from well to well.
- Raised well rims and smooth surface for reliable closing, even with heat sealing.
- Easy and reliable stacking.
- g-Safe® exceptional centrifugation stability up to 6,000 × g for faster
- protocols and improved sample quality.
- High-quality polypropylene guarantees high resistance to chemicals, mechanical stress and temperature extremes.
- Manufactured without slip agents, plasticizers, and biocides-Materials which could have a negative effect on bioassays.
- Autoclavable (121 °C, 20 min).
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service