Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

EP0030129539

Eppendorf® twin.tec(R) PCR plate 96 LoBind

skirted, with barcode(s), clear, set of 25 pieces

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106300
material:
transparent wells
size:
96 well
feature:
skirt
skirted, with barcode(s)
Product EP0030129539 is not currently sold in your country. Contact Technical Service

material

transparent wells

feature

skirt
skirted, with barcode(s)

packaging

set of 25 pieces

manufacturer/tradename

Eppendorf® 0030129539

technique(s)

PCR: suitable

size

96 well

color

clear

General description

[TM=“Eppendorf”]  [TM=“twin.tec”] PCR plate 96 [TM=“LoBind”] are specially designed PCR plates to minimize sample loss and binding of molecules. The polypropylene wells with LoBind characteristics, prevent the binding of molecules to the wells, thereby maximizing the target yield.

Application

[TM=“Eppendorf”] [TM=“twin.tec”] PCR plate 96 [TM=“LoBind”] are used extensively in:
  • PCR and real time PCR applications
  • Forensic trace analysis
  • Construction of DNA libraries for NGS workflows

Features and Benefits

  • The [TM=“LoBind”] material enhances the outcome of assays by ensuring maximum sample retrieval.
  • The plate is free from surface coatings (such as silicone) to minimize the risk of sample interference.
  • [TM=“Eppendorf”]  [TM=“twin.tec”] PCR plate 96 LoBind plates feature a distinctive [TM="OptiTrack"] matrix, enabling faster well identification and reducing pipetting errors by 30%.
  • The presence of raised well rims and a smooth surface ensure secure sealing within the plate.
  • Free of human DNA, DNase, RNase, and PCR inhibitors (PCR clean).

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service