Key Documents
EP0030128702
Eppendorf® Twin tec PCR plates, 96 wells
skirted, with barcode, def. color (wells colorless), pkg of 300 ea
About This Item
UNSPSC Code:
41106300
material
colorless wells
polycarbonate
feature
skirt
packaging
pkg of 300 ea
manufacturer/tradename
Eppendorf® 0030128702
technique(s)
PCR: suitable
size
96 wells
General description
Choose Eppendorf twin.tec PCR plates for confidence in every PCR assay. twin.tec plates offer purity, rigidity, color, and other simple, smart features can make your daily life a lot easier. The rigid polycarbonate frame works well in automation and high-throughput processes, and the different colors mean you can quickly organize the different steps of your workflow.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)