Key Documents
EP0030128338
Eppendorf® Twin tec PCR plates, 384 wells
with barcode, def. color (wells colorless), pkg of 300 ea
About This Item
UNSPSC Code:
41106300
material:
conical bottom colorless wells
size:
384 wells
Product EP0030128338 is not currently sold in your country. Contact Technical Service
material
conical bottom colorless wells
packaging
pkg of 300 ea
manufacturer/tradename
Eppendorf® 0030128338
technique(s)
PCR: suitable
size
384 wells
General description
“Eppendorf” Twin tec PCR plates with 384 wells are large scale PCR plates used in high throughput PCR workflows. They have rigid polycarbonate frames, polypropylene wells and raised well rims, ensuring a sturdy plate with effective sealing.
Application
Eppendorf “twin.tec” PCR Plates may be used in a wide variety of applications such as:
  • Standard PCR
  • Real time PCR
  • High throughput PCR
Features and Benefits
  • [TM=“Eppendorf”] Twin tec PCR plates have a unique one-piece design consisting of polypropylene wells and a polycarbonate frame.
  • Thin-walled polypropylene wells ensure optimum heat transfer to the sample.
  • The polycarbonate frame is exceptionally sturdy and torque resistant.
  • [TM=“Eppendorf”] Twin tec PCR plates display an improved well to well tolerance.
  • [TM=“Eppendorf”] Twin tec PCR plates have been batch tested and certified PCR clean.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)