EP0030125207
Eppendorf® Microtube 3810X
capacity 1.5 mL, yellow, pack of 1000 ea
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121702
material
cap (plug seal)
polypropylene cap
polypropylene tube
yellow
yellow cap
yellow tube
feature
neck thread Push fit
packaging
pack of 1000 ea
manufacturer/tradename
Eppendorf® 0030125207
capacity
1.5 mL
Looking for similar products? Visit Product Comparison Guide
General description
The easy-to-open and close design of the Eppendorf® 3810X tubes offers convenient, ergonomic and reliable one-hand operation. Whether sample preparation, centrifugation or storage, the Eppendorf® 3810X tubes fit perfectly into your workflows.
Features and Benefits
- Tube lid is easy to open and close for ergonomic operation
- Polypropylene provides high resistance to chemicals, mechanical stress, and temperature extremes g-Safe®: Exceptional centrifugation stability up to 30,000 x g prevents sample loss due to tube breakage and provides extra safety when working with hazardous samples
- Manufactured without the use of slip agents, plasticizers, and biocides – substances that have been shown to leach from plastic consumables into the sample and negatively affect bio-assay results
- Graduation for visual control of the pipetted volume
- Frosted lid and surface on the side for easy labeling
- Ensured functionality from -86 °C to 100 °C
- Autoclavable when open (121 °C, 20 min)
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
g-Safe is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service