Key Documents
EP0030079345
Eppendorf® macro Vis Cuvette
max. filling volume 4,500 μL, pkg of 1000 cuvettes (10 boxes x 100 cuvettes)
Select a Size
1 PKG
$244.00
$244.00
Check Cart for Availability
Select a Size
Change View
1 PKG
$244.00
About This Item
UNSPSC Code:
41121813
NACRES:
NB.44
$244.00
Check Cart for Availability
material
PMMA
feature
max. filling volume 4,500 μL
packaging
pkg of 1000 cuvettes (10 boxes x 100 cuvettes)
manufacturer/tradename
Eppendorf® 30079345
capacity
-4500 μL
General description
“Eppendorf” macro Vis Cuvettes are single use plastic cuvettes with a maximum volume of 4500 μL.
Application
“Eppendorf” macro Vis Cuvettes are ideal for applications that go beyond the UV range, including:
  • colorimetric protein assays like Bradford and Lowry methods
  • determining the optical density of bacterial cultures using OD 600 methods
  • as well as kinetic and fluorescence measurements.
Features and Benefits
  • Macro Vis cuvettes are made of clear plastics.
  • These cuvettes are ideal for measurements withing the wavelength range of 300 – 900 nm.
  • “Eppendorf” macro Vis Cuvettes are specifically used in controlling temperature for kinetic measurements.
  • Suitable for use with Eppendorf BioSpectrometer fluorescence and Eppendorf BioPhotometer
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)