EP0030077806
Eppendorf® ep Dualfilter T.I.P.S.® SealMax G
volume (0.1-10 μL), PCR clean, sterile, Pyrogen free, pack of 960 ea (10 racks of 96 tips)
About This Item
Recommended Products
material
gray
sterility
sterile
feature
DNase free
PCR clean
RNase free
filter
Pyrogen free
packaging
pack of 960 ea (10 racks of 96 tips)
manufacturer/tradename
Eppendorf® 0030077806
technique(s)
PCR: suitable
maximum volume
10 μL
volume
(0.1-10 μL)
General description
ep Dualfilter T.I.P.S. and ep Dualfilter T.I.P.S. SealMax are manufactured to the highest quality standards from pure, non-recycled materials under cleanroom conditions without the use of interfering slip agents, biocides or plasticizers.
Ideally suited for methods where high sensitivity and reproducibility are essential, both ep Dualfilter T.I.P.S. and ep Dualfilter T.I.P.S. SealMax are especially recommended for applications such as PCR and microbiology where preserving sample integrity and preventing cross contamination are critical.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)