Key Documents
EP0030077750
Eppendorf® ep Dualfilter T.I.P.S.®
5 racks x 96 ea, PCR clean, pyrogen free, capacity 50-1250 μL, sterile
About This Item
UNSPSC Code:
41121600
volume:
50—1250 μL
sterility:
sterile
feature:
filter
packaging:
pack of 480 ea (5 racks of 96 tips)
manufacturer/tradename:
Eppendorf® 0030077750
Product EP0030077750 is not currently sold in your country. Contact Technical Service
material
green
sterility
sterile
feature
filter
packaging
pack of 480 ea (5 racks of 96 tips)
manufacturer/tradename
Eppendorf® 0030077750
technique(s)
PCR: suitable
capacity
50-1250 μL
maximum volume
1.25 mL
volume
50—1250 μL
General description
  • Eppendorf® ep Dualfilter T.I.P.S.® of 50-1250 μL capacity are well suited for applications with samples that contain detergents.
  • Ultrahomogenous surface for maximum reproducibility.
  • Ultrahydrophobic surface for minimal sample loss.
  • Significantly reduced foam formation during pipetting.
  • Extreme resistance to chemicals.
  • Available in PCR clean and Eppendorf Quality.
  • Also available as ep Dualfilter T.I.P.S. in PCR clean/Sterile (sterile and pyrogen-free).
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
ep Dualfilter T.I.P.S. is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)