Key Documents
EP0030077741
Eppendorf® ep Dualfilter T.I.P.S.® LoRetention
tip volume × L 2-200 μL × 55 mm, PCR clean, sterile, case of 960 tips 10 racks × 96 tips
About This Item
UNSPSC Code:
41121600
material
yellow tip
sterility
sterile
feature
PCR clean
packaging
case of 960 ea
case of 960 tips 10 racks × 96 tips
manufacturer/tradename
Eppendorf® 0030077741
tip volume × L
2-200 μL × 55 mm
General description
  • Ultrahomogenous surface for maximized reproducibility
  • Ultrahydrophobic surface for minimal sample loss
  • Significantly reduced foam formation during pipetting
  • Extreme resistance to chemicals
  • Available in PCR clean and Eppendorf Quality
  • Also available as ep Dualfilter T.I.P.S. in PCR clean/Sterile (sterile and pyrogen-free)
  • No coating
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
ep Dualfilter T.I.P.S. is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)