Key Documents
EP0030077636
Eppendorf® ep Dualfilter T.I.P.S.®
10 trays x 96 ea, PCR clean, capacity 20-300 μL, sterile
About This Item
UNSPSC Code:
41121600
volume:
20—300 μL
sterility:
sterile
feature:
DNase free
RNase free
filter
packaging:
pack of 960 ea (10 trays of 96 tips)
manufacturer/tradename:
Eppendorf® 0030077636
Product EP0030077636 is not currently sold in your country. Contact Technical Service
material
orange
sterility
sterile
feature
DNase free
RNase free
filter
packaging
pack of 960 ea (10 trays of 96 tips)
manufacturer/tradename
Eppendorf® 0030077636
technique(s)
PCR: suitable
capacity
20-300 μL
maximum volume
300 μL
volume
20—300 μL
General description
  • Eppendorf® ep Dualfilter T.I.P.S.® of 20-300 μL capacity are uncoated, free of additives and do not leach into the sample.
  • Ultrahomogenous surface for maximum reproducibility.
  • Ultrahydrophobic surface for minimal sample loss.
  • Significantly reduced foam formation during pipetting.
  • Extreme resistance to chemicals.
  • Available in PCR clean and Eppendorf Quality.
  • Also available as ep Dualfilter T.I.P.S. in PCR clean/Sterile (sterile and pyrogen-free).
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
ep Dualfilter T.I.P.S. is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)