Key Documents
EP0030077628
Eppendorf® ep Dualfilter T.I.P.S.®
10 trays x 96 ea, PCR clean, capacity 0.5-20 μL, sterile
About This Item
UNSPSC Code:
41121600
volume:
0.5—20 μL
sterility:
sterile
feature:
DNase free
RNase free
filter
packaging:
pack of 960 ea (10 trays of 96 tips)
manufacturer/tradename:
Eppendorf® 0030077628
Product EP0030077628 is not currently sold in your country. Contact Technical Service
material
gray
sterility
sterile
feature
DNase free
RNase free
filter
packaging
pack of 960 ea (10 trays of 96 tips)
manufacturer/tradename
Eppendorf® 0030077628
technique(s)
PCR: suitable
capacity
0.5-20 μL
maximum volume
20 μL
volume
0.5—20 μL
General description
  • Eppendorf® ep Dualfilter T.I.P.S.® of 0.5-20 μL capacity are available as PCR clean and Eppendorf quality.
  • They are uncoated, free of additives and do not leach into the sample.
  • Ultrahomogenous surface for maximum reproducibility.
  • Ultrahydrophobic surface for minimal sample loss.
  • Significantly reduced foam formation during pipetting.
  • Extreme resistance to chemicals.
  • Available in PCR clean and Eppendorf Quality.
  • Also available as ep Dualfilter T.I.P.S. in PCR clean/Sterile (sterile and pyrogen-free).
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
ep Dualfilter T.I.P.S. is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)