Key Documents
EP0030077610
Eppendorf® ep Dualfilter T.I.P.S.®
10 trays x 96 ea, PCR clean, capacity 0.1-10 μL, sterile
About This Item
UNSPSC Code:
41121600
volume:
0.1—10 μL
sterility:
sterile
feature:
DNase free
RNase free
filter
packaging:
pack of 960 ea (10 trays of 96 tips)
manufacturer/tradename:
Eppendorf® 0030077610
Product EP0030077610 is not currently sold in your country. Contact Technical Service
material
gray
sterility
sterile
feature
DNase free
RNase free
filter
packaging
pack of 960 ea (10 trays of 96 tips)
manufacturer/tradename
Eppendorf® 0030077610
technique(s)
PCR: suitable
capacity
0.1-10 μL
maximum volume
10 μL
volume
0.1—10 μL
General description
  • Eppendorf® ep Dualfilter T.I.P.S.® of 0.1-10 μL capacity are generated by an ultrahydrophobic, extremely homogeneous surface for maximum reproducibility and minimum sample loss.
  • Significantly reduced foam formation during pipetting.
  • Extreme resistance to chemicals.
  • Available in PCR clean and Eppendorf Quality.
  • Also available as ep Dualfilter T.I.P.S. in PCR clean/Sterile (sterile and pyrogen-free).
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
ep Dualfilter T.I.P.S. is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)