Key Documents
EP0030077580
Eppendorf® ep Dualfilter T.I.P.S.®
5 racks x 24 ea, PCR clean, capacity 100-5000 μL, sterile
About This Item
UNSPSC Code:
41121600
volume:
100—5000 μL
sterility:
sterile
feature:
DNase free
RNase free
filter
packaging:
pack of 120 ea (5 racks of 24 tips)
manufacturer/tradename:
Eppendorf® 0030077580
sterility
sterile
feature
DNase free
RNase free
filter
packaging
pack of 120 ea (5 racks of 24 tips)
manufacturer/tradename
Eppendorf® 0030077580
technique(s)
PCR: suitable
capacity
100-5000 μL
maximum volume
5 mL
volume
100—5000 μL
General description
  • Eppendorf® ep Dualfilter T.I.P.S.® of 100-5000 μL capacity are well suited for applications with samples that contain detergents.
  • Two filter layers provide dual protection against aerosol and biomolecules to guard your pipette and sample.
  • Available as PCR clean/Sterile (sterile and pyrogen-free).
  • Continuous inspection of each production lot by an independent laboratory guarantees high purity and quality.
  • Ultrahomogenous surface for maximum reproducibility.
  • Ultrahydrophobic surface for minimal sample loss.
  • Significantly reduced foam formation during pipetting.
  • Extreme resistance to chemicals.
  • Available in PCR clean and Eppendorf Quality.
  • Also available as ep Dualfilter T.I.P.S. in PCR clean/Sterile.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
ep Dualfilter T.I.P.S. is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)