Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EP0030077555

Eppendorf® ep Dualfilter T.I.P.S.®

10 racks x 96 ea, PCR clean, capacity 2-200 μL, sterile

Synonym(s):

pipetting tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121600
volume:
2—200 μL
sterility:
sterile
feature:
DNase free
RNase free
filter
packaging:
pack of 960 ea (10 racks of 96 tips)
manufacturer/tradename:
Eppendorf® 0030077555
Product EP0030077555 is not currently sold in your country. Contact Technical Service

material

yellow (Tray)

sterility

sterile

feature

DNase free
RNase free
filter

packaging

pack of 960 ea (10 racks of 96 tips)

manufacturer/tradename

Eppendorf® 0030077555

technique(s)

PCR: suitable

capacity

2-200 μL

maximum volume

200 μL

volume

2—200 μL

Looking for similar products? Visit Product Comparison Guide

General description

  • Eppendorf® ep Dualfilter T.I.P.S.® of 2-200 μL capacity are well suited for applications with samples that contain detergents.
  • Two filter layers provide dual protection against aerosol and biomolecules to guard your pipette and sample.
  • Available as PCR clean/Sterile (sterile and pyrogen-free).
  • Continuous inspection of each production lot by an independent laboratory guarantees high purity and quality.
  • Ultrahomogenous surface for maximum reproducibility.
  • Ultrahydrophobic surface for minimal sample loss.
  • Significantly reduced foam formation during pipetting.
  • Extreme resistance to chemicals.
  • Available in PCR clean and Eppendorf Quality.
  • Also available as ep Dualfilter T.I.P.S. in PCR clean/Sterile.

Legal Information

Eppendorf is a registered trademark of Eppendorf AG
ep Dualfilter T.I.P.S. is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service