Key Documents
EP0030077520
Eppendorf® ep Dualfilter T.I.P.S.®
10 racks x 96 ea, PCR clean, capacity 0.5-20 μL, sterile
About This Item
UNSPSC Code:
41121600
volume:
0.5—20 μL
sterility:
sterile
feature:
DNase free
RNase free
filter
packaging:
pack of 960 ea (10 racks of 96 tips)
manufacturer/tradename:
Eppendorf® 0030077520
Product EP0030077520 is not currently sold in your country. Contact Technical Service
material
gray (Tray)
sterility
sterile
feature
DNase free
RNase free
filter
packaging
pack of 960 ea (10 racks of 96 tips)
manufacturer/tradename
Eppendorf® 0030077520
technique(s)
PCR: suitable
capacity
0.5-20 μL
maximum volume
20 μL
volume
0.5—20 μL
General description
  • Eppendorf® ep Dualfilter T.I.P.S.® (0.5-20 μL) have low retention surfaces, which increases the reproducibility and significantly reduce the loss of expensive reagents.
  • Ultrahomogenous surface for maximum reproducibility.
  • Ultrahydrophobic surface for minimal sample loss.
  • Significantly reduced foam formation during pipetting.
  • Extreme resistance to chemicals.
  • Available in PCR clean and Eppendorf Quality.
  • Also available as ep Dualfilter T.I.P.S. in PCR clean/Sterile.
  • Sterile and pyrogen-free.
  • No Coating.
Application
  • Cell culture (media).
  • Genomics: PCR, RT-PCR, qPCR and all other types of PCR.
  • Enzymatic reactions (restriction decomposition, ligation).
  • Isolation and purification of nucleic acid.
  • Gel electrophoresis (e.g., prefabricated DNA ladders) typical detergents: SDS, Triton, X-100, Brij® 35, Tween® 20, CHAPS.
  • Proteomics (all types of protein examinations).
  • Protein isolation and purification.
  • NGS library preparation.
Legal Information
Brij is a registered trademark of Croda International PLC
Eppendorf is a registered trademark of Eppendorf AG
TWEEN is a registered trademark of Croda International PLC
Triton is a trademark of The Dow Chemical Company or an affiliated company of Dow
ep Dualfilter T.I.P.S. is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)