Key Documents
EP0030075137
Eppendorf® epT.I.P.S. Biopur®, racks
volume × L 0.1-5.0 mL × 120 mm, violet, pkg of 120 tips (5 racks × 24 tips)
About This Item
UNSPSC Code:
41121600
material
violet
sterility
sterile
feature
Eppendorf Biopur®
packaging
pkg of 120 tips (5 racks × 24 tips)
manufacturer/tradename
Eppendorf® 0030075137 (EU)
volume × L
0.1-5.0 mL × 120 mm
color
violet
foreign activity
PCR inhibitors, human and baterial DNA, DNase, RNase, ATP, Pyrogen, free
General description
Biopur guaranteed sterile, pyrogen-free, RNase-free, DNA-free and ATP-free, for applications in pharmaceutical and food industries, molecular biology and cell technology.
Batch specific certificates available on the internet at http://www.eppendorf.com
Legal Information
Biopur is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)