Key Documents
EP0030072049
Eppendorf® epT.I.P.S. LoRetention reload
capacity 0.1-10 μL , S, 10 trays x 96 ea
About This Item
UNSPSC Code:
41121600
NACRES:
NB.01
Product EP0030072049 is not currently sold in your country. Contact Technical Service
material
gray (Tray)
sterility
non-sterile
packaging
pack of 960 ea (Trays)
manufacturer/tradename
Eppendorf® 0030072049
capacity
0.1-10 μL , S
maximum volume
10 μL
General description
  • Eppendorf® epT.I.P.S. LoRetention pipette tips reloads (dark gray) are particularly well-suited for applications with samples that contain detergents. Detergents or detergent containing liquids lower the liquid′s surface tension. This leads to a phenomenon called “wetting”: Residual liquid sticks to the tip wall and cannot be recovered.
  • The invisible film left behind in standard tips shows a significantly high amount of valuable sample material which becomes discarded together with the tip. Particularly when it comes to sensitive PCR and real-time PCR applications or NGS library preparation, low retention surfaces can increase the reproducibility and significantly reduce the loss of expensive reagents.
  • The low retention property of epT.I.P.S. LoRetention pipette tips is generated by an ultrahydrophobic, extremely homogeneous surface. It is achieved through an innovative treatment at the molecular level − the “Pearl Effect” Technology.
  • All pipette tips are uncoated, free of additives and do not leach into the sample.
  • Liquids containing detergents simply roll off, so that practically no liquid remains in the tip.
  • These outstanding material properties ensure maximized sample recovery for cost saving and improved reproducibility.
Application
  • Cell culture (media).
  • Genomics: PCR, RT-PCR, qPCR and all other types of PCR.
  • Enzymatic reactions (restriction decomposition, ligation).
  • Isolation and purification of nucleic acid.
  • Gel electrophoresis (e.g., prefabricated DNA ladders) typical detergents: SDS, Triton, X-100, Brij® 35, Tween® 20, CHAPS.
  • Proteomics (all types of protein examinations).
  • Protein isolation and purification, NGS library preparation.
Features and Benefits
  • Ultrahomogenous surface for maximum reproducibility.
  • Ultrahydrophobic surface for minimal sample loss.
  • Significantly reduced foam formation during pipetting.
  • Extreme resistance to chemicals.
  • No coating.
Legal Information
Brij is a registered trademark of Croda International PLC
Eppendorf is a registered trademark of Eppendorf AG
TWEEN is a registered trademark of Croda International PLC
Triton is a trademark of The Dow Chemical Company or an affiliated company of Dow
Choose from one of the most recent versions:
Certificates of Analysis (COA)