Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EP0030072022

Eppendorf® epT.I.P.S. LoRetention reload

capacity 2-200 μL, 10 trays x 96 ea, PCR clean

Synonym(s):

pipetting tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121600
Product EP0030072022 is not currently sold in your country. Contact Technical Service

material

yellow (Tray)

sterility

non-sterile

feature

DNase free
RNase free

packaging

pack of 960 ea (Trays)

manufacturer/tradename

Eppendorf® 0030072022

technique(s)

PCR: suitable

capacity

2-200 μL

maximum volume

200 μL

Looking for similar products? Visit Product Comparison Guide

General description

  • Eppendorf® epT.I.P.S. LoRetention pipette tips reloads (yellow) are particularly well-suited for applications with samples that contain detergents.
  • Detergents or detergent containing liquids lower the liquid′s surface tension. This leads to a phenomenon called “wetting”: Residual liquid sticks to the tip wall and cannot be recovered.
  • The invisible film left behind in standard tips shows a significantly high amount of valuable sample material which becomes discarded together with the tip. Particularly when it comes to sensitive PCR and real-time PCR applications or NGS library preparation, low retention surfaces can increase the reproducibility and significantly reduce the loss of expensive reagents.
  • The low retention property of epT.I.P.S. LoRetention pipette tips is generated by an ultrahydrophobic, extremely homogeneous surface. It is achieved through an innovative treatment at the molecular level − the “Pearl Effect” Technology.
  • All pipette tips are uncoated, free of additives and do not leach into the sample.
  • Liquids containing detergents simply roll off, so that practically no liquid remains in the tip.
  • These outstanding material properties ensure maximized sample recovery for cost saving and improved reproducibility.

Application

  • Cell culture (media).
  • Genomics: PCR, RT-PCR, qPCR and all other types of PCR.
  • Enzymatic reactions (restriction decomposition, ligation).
  • Isolation and purification of nucleic acid.
  • Gel electrophoresis (e.g., prefabricated DNA ladders) typical detergents: SDS, Triton, X-100, Brij® 35, Tween® 20, CHAPS.
  • Proteomics (all types of protein examinations).
  • Protein isolation and purification, NGS library preparation.

Features and Benefits

  • Significantly reduced foam formation during pipetting.
  • Extreme resistance to chemicals.
  • Available in PCR clean.
  • No coating.

Legal Information

Brij is a registered trademark of Croda International PLC
Eppendorf is a registered trademark of Eppendorf AG
TWEEN is a registered trademark of Croda International PLC
Triton is a trademark of The Dow Chemical Company or an affiliated company of Dow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service