Key Documents
EP0030050568
Eppendorf® Maxitip for Varitip S
pkg of 200 ea
About This Item
UNSPSC Code:
41121600
NACRES:
NB.02
sterility:
non-sterile
feature:
graduations
packaging:
pack of 200 ea
pkg of 200 ea
manufacturer/tradename:
Eppendorf® 0030050568
Product EP0030050568 is not currently sold in your country. Contact Technical Service
sterility
non-sterile
feature
graduations
packaging
pack of 200 ea
pkg of 200 ea
manufacturer/tradename
Eppendorf® 0030050568
General description
Eppendorf VaritipS are used in combination with the Eppendorf Maxitip to aspirate between 2.5 mL and 10 mL of liquid from tall, narrow-necked vessels using the air cushion principle. The Varitips valve ensures drip-free dosing of liquids with high vapor pressure Varitips also have graduations ensuring accurate dosing of non-aqueous liquids.
Features and Benefits
  • Use Varitips P for aspirating 1 mL to 10 mL from beakers using the positive displacement principle
  • Use Varitips S in combination with the Eppendorf Maxitip to aspirate between 2.5 mL and 10 mL of liquid from tall, narrow-necked vessels using the air cushion principle
  • Valve for Varitips ensures drip-free dosing of liquids with high vapor pressure
  • The graduation on Varitips ensures accurate dosing of non-aqueous liquids
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)