Key Documents
EP0030050541
Eppendorf® Valve for Varitips S
pkg of 100 pieces
About This Item
UNSPSC Code:
41121600
NACRES:
NB.02
Product EP0030050541 is not currently sold in your country. Contact Technical Service
packaging
pkg of 100 pieces
manufacturer/tradename
0030050541
Eppendorf®
General description
Varitips S are used in combination with the Eppendorf Maxitip to aspirate between 2.5 mL and 10 mL of liquid from tall, narrow-necked vessels using the air cushion principle.
Features and Benefits
  • Use Varitips P for aspirating 1 mL to 10 mL from beakers using the positive displacement principle
  • Use Varitips S in combination with the Eppendorf Maxitip to aspirate between 2.5 mL and 10 mL of liquid from tall, narrow-necked vessels using the air cushion principle
  • Valve for Varitips ensures drip-free dosing of liquids with high vapor pressure
  • The graduation on Varitips ensures accurate dosing of non-aqueous liquids
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)