Key Documents
EP0030050533
Eppendorf® Dispensing Part
for Varitip S, pkg of 30 pieces
About This Item
UNSPSC Code:
41121500
NACRES:
NB.51
Product EP0030050533 is not currently sold in your country. Contact Technical Service
feature
for Varitip S
packaging
pkg of 30 pieces
manufacturer/tradename
0030050533
Eppendorf®
General description
The Eppendorf Varitips S dosing unit is a replacement dispensing part for use with Maxitips and consists of a barrel with a piston. The Varipette/Maxipettor becomes a functional unit when a Varitip is attached. The Varitip consists of a piston, barrel and tip and holds the liquid to be dispensed.
Features and Benefits
  • Use Varitips P for aspirating 1 mL to 10 mL from beakers using the positive displacement principle
  • Use Varitips S in combination with the Eppendorf Maxitip to aspirate between 2.5 mL and 10 mL of liquid from tall, narrow-necked vessels using the air cushion principle
  • Valve for Varitips ensures drip-free dosing of liquids with high vapor pressure
  • The graduation on Varitips ensures accurate dosing of non-aqueous liquids
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)