Key Documents
EP0030001320
Eppendorf® Mastertips, tips plus piston, ready-for-use
pack of 480 ea
About This Item
UNSPSC Code:
41121600
material
gray
sterility
non-sterile
packaging
pack of 480 ea
manufacturer/tradename
Eppendorf® 0030001320
maximum volume
20 μL
General description
Eppendorf® Mastertips, tips plus piston (light gray), ready-for-use with the integrated piston are syringe-style, positive displacement tips to prevent pipette contamination and guarantee accurate pipetting of viscous solutions or liquids with high-vapor pressure.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)