Key Documents
EP0030001222
Eppendorf® epT.I.P.S. GELoader, 2 racks x 96 ea
2 racks x 96 ea
About This Item
UNSPSC Code:
41121600
material
gray
sterility
non-sterile
packaging
pack of 96 ea (Racked)
manufacturer/tradename
Eppendorf® 0030001222
maximum volume
20 μL
General description
  • Special tip for gel electrophoresis, loading of polyacrylamide gels.
  • Flexible long and narrow tips for handling of smallest volumes.
  • GELoader tips and rack autoclavable.
  • Highest precision and accuracy when used with Eppendorf pipettes for 0.5 to 10 μL (gray control button).
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)