Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ELDEXACID

Sigma-Aldrich

Elphodex Acidic Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352201

technique(s)

electrophoresis: suitable

shipped in

wet ice

storage temp.

2-8°C

Application

Cyclodextrins are widely used in capillary electrophoresis for complexation and separation of enantiomers. Cyclodextrins are also used to modify the mobility of isomers and structurally similar compounds, allowing for more efficient separation by capillary electrophoresis and HPLC. Sigma-Aldrich provides these sets of cyclodextrins to allow the researcher to develop and optimize their chiral CE systems.
The Elphodextrin Starter Kit is designed for overall capillary electrophoresis system development. The Elphodex A, B and G kits include modified forms of α−, β−, or γ−cyclodextrin for further optimization after determination of the ideal pore size for separation. The Elphodex Acid and Elphodex Phosphate kits include α−, β−, and γ−cyclodextrins modified to create an ionic charge on the cyclodextrin molecule and are used for separation of neutral compounds.

Other Notes

Contains α-, β- and γ- cyclodextrins modified with carboxymethyl or carboxyethyl groups to create an ionic charge on the cyclodextrin molecule. For optimization of CE separation of neutral compounds.

Kit Components Only

Product No.
Description

  • Carboxymethyl α-cyclodextrin .5 g

  • Carboxymethyl β-cyclodextrin .5 g

  • Carboxymethyl γ-cyclodextrin .5 g

  • Carboxyethyl α-cyclodextrin .5 g

  • Carboxyethyl β-cyclodextrin .5 g

  • Carboxyethyl γ-cyclodextrin .5 g


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service