Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EHU106481

Sigma-Aldrich

MISSION® esiRNA

targeting human TRIM16, RP11-385D13.1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

AGACTTGGAGCGGAAACTCAAGTTGAATGAAAATGCCATCTCCAGGCTCCAGGCTAACCAAAAGTCTGTTCTGGTGTCGGTGTCAGAGGTCAAAGCGGTGGCTGAAATGCAGTTTGGGGAACTCCTTGCTGCTGTGAGGAAGGCCCAGGCCAATGTGATGCTCTTCTTAGAGGAGAAGGAGCAAGCTGCGCTGAGCCAGGCCAACGGTATCAAGGCCCACCTGGAGTACAGGAGTGCCGAGATGGAGAAGAGCAAGCAGGAGCTGGAGAGGATGGCGGCCATCAGCAACACTGTCCAGTTCTTGGAGGAGTACTGCAAGTTTAAGAACACTGAAGACATCACCTTCCCTAGTGTTTACGTAGGGCTGAAGGATAAACTCTCGGGCATCCGCAAAGTTATCACGGAATCCACTGTACACTTAATCCAGTTGCTGGAGAACTATAAGAAAAAGCTCCAGGAGTTTTCCAAGGAAGAGGAGTATGACATCAGAACTCAAGTGTCTGCCGTTGTTC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zsuzsanna Nagy et al.
Scientific reports, 10(1), 19724-19724 (2020-11-14)
Although selective BRAF inhibitors and novel immunotherapies have improved short-term treatment responses in metastatic melanoma patients, acquired resistance to these therapeutics still represent a major challenge in clinical practice. In this study, we evaluated the efficacy of Withaferin A (WFA)
Yi Zhao et al.
Frontiers in cell and developmental biology, 8, 625105-625105 (2021-01-26)
Bone regeneration is the ultimate goal of periodontal therapies, in which osteogenic differentiation of human periodontal ligament stem cells plays a critical role. The tripartite motif (TRIM)16, an E3 ubiquitin ligase, is downregulated in periodontal tissues of patients with periodontitis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service