Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EHU091601

Sigma-Aldrich

MISSION® esiRNA

targeting human DNAJA3

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$220.00
50 μG
$391.00

$220.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
20 μG
$220.00
50 μG
$391.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$220.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TGGTGTCAGCCTTACAGGAACAAAACATAACCCTTTCATTTGTACTGCCTCCTTCCACACGAGTGCCCCTTTGGCCAAAGAAGATTATTATCAGATATTAGGAGTGCCTCGAAATGCCAGCCAGAAAGAGATCAAGAAAGCCTATTATCAGCTTGCCAAGAAGTATCACCCTGACACAAATAAGGATGATCCCAAAGCCAAGGAGAAGTTCTCCCAGCTGGCAGAAGCCTATGAGGTTTTGAGTGATGAGGTGAAGAGGAAGCAGTACGATGCCTACGGCTCTGCAGGCTTCGATCCTGGGGCCAGCGGCTCCCAGCATAGCTACTGGAAGGGAGGCCCCACTGTGGACCCCGAGGAGCTGTTCAGGAAGATCTTTGGCGAGTTCTCATCCTCTTCATTTGGAGATTTCCAGACCGTGTTTGATCA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kohei Kumada et al.
Biochemical and biophysical research communications, 513(4), 947-951 (2019-04-22)
Nuclear factor kappa B (NF-κB) controls the transcription of various genes in response to immune stimuli. Our previous study revealed that the Droj2/DNAJA3 cochaperone contributes to the NF-κB pathway in Drosophila and humans. In general, the cochaperone is associated with
Sheng-Fan Wang et al.
Cancers, 12(11) (2020-11-26)
Gastric cancer is a common health issue. Deregulated cellular energetics is regarded as a cancer hallmark and mitochondrial dysfunction might contribute to cancer progression. Tid1, a mitochondrial co-chaperone, may play a role as a tumor suppressor in various cancers, but

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service