Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EHU084591

Sigma-Aldrich

MISSION® esiRNA

targeting human MFGE8

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$220.00
50 μG
$391.00

$220.00


Usually ships in 3 weeks (4 to 6 weeks for custom orders).


Select a Size

Change View
20 μG
$220.00
50 μG
$391.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$220.00


Usually ships in 3 weeks (4 to 6 weeks for custom orders).

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

AGGAGATGTCTTCCCCTCGTACACCTGCACGTGCCTTAAGGGCTACGCGGGCAACCACTGTGAGACGAAATGTGTCGAGCCACTGGGCCTGGAGAATGGGAACATTGCCAACTCACAGATCGCCGCCTCGTCTGTGCGTGTGACCTTCTTGGGTTTGCAGCATTGGGTCCCGGAGCTGGCCCGCCTGAACCGCGCAGGCATGGTCAATGCCTGGACACCCAGCAGCAATGACGATAACCCCTGGATCCAGGTGAACCTGCTGCGGAGGATGTGGGTAACAGGTGTGGTGACGCAGGGTGCCAGCCGCTTGGCCAGTCATGAGTACCTGAAGGCCTTCAAGGTGGCCTACAGCCTTAATGGACACGAATTCGATTTCATCCATGATGTTAATAAAAAACACAAGGAGTTTGTGGGTAACTGGAACAAAAACGCGG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yong-Yue Gao et al.
Cell death & disease, 9(9), 845-845 (2018-08-30)
Accumulating evidence suggests neuronal apoptosis has the potential to lead to more harmful effects in the pathological processes following traumatic brain injury (TBI). Previous studies have established that milk fat globule-EGF factor-8 (MFG-E8) provides neuroprotection through modulation of inflammation, oxidative
Yong-Yue Gao et al.
Experimental neurology, 336, 113532-113532 (2020-11-28)
Increasing evidence suggests that microglial polarization plays an important role in the pathological processes of neuroinflammation following subarachnoid hemorrhage (SAH). Previous studies indicated that milk fat globule-epidermal growth factor-8 (MFG-E8) has potential anti-apoptotic and anti-inflammatory effects in cerebral ischemia. However
A Ellert-Miklaszewska et al.
Oncogene, 35(50), 6366-6377 (2016-04-05)
Tumour tissue is infiltrated by myeloid cells that are reprogrammed into alternatively activated/regenerative (M2) macrophages. The contribution of major signalling pathways and their modulators/targets involved in the macrophage reprogramming is poorly known. Glioblastoma (malignant brain tumour) attracts and reprograms brain-resident

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service