Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

E9654

Sigma-Aldrich

S1P4 C-Terminal Blocking Peptide

~1 mg/mL, buffered aqueous solution

Synonym(s):

EDG-6, Endothelial Cell Differentiation Gene 6

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352211
NACRES:
NA.32

form

buffered aqueous solution

Quality Level

mol wt

~2.5 kDa

concentration

~1 mg/mL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... S1PR4(8698)

General description

S1PR4 (sphingosine-1-phosphate receptor 4) is a member of the S1P or the Edg (endothelial differentiation gene) receptor family, which contains five members. It is a G-protein coupled receptor (GPCR). Its 3rd transmembrane domain contains a conserved glutamic acid residue.[1] It is expressed in lung, hematopoietic and lymphoid tissue.[2] This gene is localized to human chromosome 19p13.3.[3]

Biochem/physiol Actions

G protein-coupled receptor whose ligand might be a lysophospholipid or lysosphingo-lipid. Specifically expressed in fetal and adult lymphoid, hematopoietic and lung tissue.
S1PR4 (sphingosine-1-phosphate receptor 4) functions as a receptor for S1P, which is a phospholipid growth factor, and might control the transport of dendritic cells (DCs). Thus, it might promote Th2 lymphocyte-mediated immunity. S1P4 mediates RAS(rat sarcoma)-MAPK (mitogen-activated protein kinase) pathway by interacting with Gi/o proteins.[4] In ER (estrogen receptor)- breast cancer, its elevated expression is linked with decreased overall survival as well as disease-free survival.[5]

Physical form

Solution in phosphate buffered saline, pH 7.3.

Analysis Note

Used as a blocking peptide in immunoblotting applications with Anti-S1P4 polyclonal and monoclonal antibodies (Prod. Nos. E 7653, E 9529).

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marco Idzko et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 16(6), 625-627 (2002-03-29)
Sphingosine 1-phosphate (S1P) is a potent extracellular lysolipid phosphoric acid mediator that is released after IgE-stimulation of mast cells. Here we investigated the biological activity and intracellular signaling of S1P on human dendritic cells (DC), which are specialized antigen presenting
M H Gräler et al.
Genomics, 53(2), 164-169 (1998-10-29)
EDG receptors constitute a novel subfamily of G-protein-coupled receptors displaying a heterogeneous expression pattern. It was shown recently that the four members of this family thus far identified can bind lysophospholipids or lysosphingolipids as ligands. Here we report the identification
J Ohotski et al.
British journal of cancer, 106(8), 1453-1459 (2012-03-31)
We previously reported that sphingosine 1-phosphate receptor 4 (S1P(4)) is expressed and stimulates the ERK-1/2 pathway via a human epidermal growth factor receptor 2 (HER2)-dependent mechanism in oestrogen receptor-negative (ER(-)) MDA-MB-453 breast cancer cells. Clinical relevance of S1P(4) and sphingosine
Gill Holdsworth et al.
BMC biochemistry, 5, 12-12 (2004-08-10)
Sphingosine-1-phosphate and lysophosphatidic acid (LPA) are ligands for two related families of G protein-coupled receptors, the S1P and LPA receptors, respectively. The lysophospholipid ligands of these receptors are structurally similar, however recognition of these lipids by these receptors is highly
J R Van Brocklyn et al.
Blood, 95(8), 2624-2629 (2001-02-07)
EDG-6 is a recently cloned member of the endothelial differentiation gene (EDG) G protein-coupled receptor family that is expressed in lymphoid and hematopoietic tissue and in the lung. Homology of EDG-6 to the known sphingosine-1-phosphate (SPP) receptors EDG-1, EDG-3, and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service