Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

E9529

Sigma-Aldrich

Monoclonal Anti-S1P4, C-Terminal antibody produced in mouse

~1 mg/mL, clone 1, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-EDG-6, Anti-Endothelial Cell Differentiating Gene 6

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1, monoclonal

form

buffered aqueous solution

species reactivity

human

concentration

~1 mg/mL

technique(s)

flow cytometry: suitable
western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... S1PR4(8698)

Specificity

Specifically detects human recombinant and native EDG-6/S1P4 protein (approximately 45 kDa).

Immunogen

synthetic peptide derived from the C-terminal region of the EDG-6/S1P4 protein.

Application

Anti-S1P4, C-Terminal antibody produced in rabbit is suitable for western blotting and flow cytometry at a working antibody concentration of 5-10μg/mL using RH7777 rat hepatoma cells transfected with EDG-6 protein.

Biochem/physiol Actions

Sphingosine-1-phosphate receptor 4 (S1PR4, EDG-6) is a human gene, which encodes a G protein-coupled receptor and is identified as an orphan G-protein-coupled receptor. It binds the lipid signaling molecule sphingosine 1-phosphate (S1P). EDG-6 is a member of the endothelial differentiation gene (EDG) G protein-coupled receptor family. It is expressed in lymphoid, hematopoietic tissue and in the lung. EDG-6 acts as a high affinity receptor for SPP that couples to a G(i/o) protein leading to the activation of growth-related signaling pathways. It is one of the S1P or lysosphingolipid receptors that couples to phospholipase C-Ca2+ system through pertussis toxin-sensitive G-proteins.

Analysis Note

To detect native EDG-6/S1P4 in cells and tissues, use higher antibody concentrations.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Yamazaki et al.
Biochemical and biophysical research communications, 268(2), 583-589 (2000-02-19)
The endothelial differentiation gene-6 (Edg-6) was recently identified as an orphan G-protein-coupled receptor. Its predicted amino acid sequence is very close to Edg family of receptor proteins whose ligand is supposed to be lysophosphatidic acid (LPA) or lysosphingolipid such as
J R Van Brocklyn et al.
Blood, 95(8), 2624-2629 (2001-02-07)
EDG-6 is a recently cloned member of the endothelial differentiation gene (EDG) G protein-coupled receptor family that is expressed in lymphoid and hematopoietic tissue and in the lung. Homology of EDG-6 to the known sphingosine-1-phosphate (SPP) receptors EDG-1, EDG-3, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service