Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

E9413

Sigma-Aldrich

Cytochrome P450 2D6 human

Synonym(s):

Cytochrome P450 Microsome Preparation human, Cerosomes

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
UNSPSC Code:
12352204

biological source

human

Quality Level

recombinant

expressed in Saccharomyces cerevisiae

form

liquid

specific activity

≥10 units/pmol enzyme

secondary activity

≥100 units/mg protein (Reductase activity)

concentration

≥10 mg/mL protein (Bradford)
1 pmol/μL

UniProt accession no.

application(s)

cell analysis

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... CYP2D6(1565)

General description

Microsome contents of CYP2D6 human and yeast CYP-reductase expressed in Saccharomyces cerevisiae.

Application

Recombinant cytochrome P450, isozyme 2D6, (CYP2D6) may be used in toxicity and drug metabolism/ pharmacokinetics research to identify its specificity and activity versus xenobiotic and endogenous compounds. Recombinant cytochrome P450, isozyme 2D6, may be used separately or as a component of a P450 isozyme panel to identify and characterize natural and synthetic selective inhibitors that affect the metabolism of toxic compounds or therapeutic drugs such as fluoxetine, codeine, and tamoxifen.

Unit Definition

P450 Activity: One unit will oxidize 1 picomole of dextromethorphan per minute at pH 7.4 at 28 °C.
Reductase Activity: One unit will reduce 1 nanomole of cytochrome C per minute in the presence of NADPH at pH 7.4 at 28 °C.

Physical form

50 mM Tris (pH 7.4), 1 mM EDTA, 20% glycerol

Other Notes

For more more information visit our drug metabolism resources for Cytochrome P450 microsome preparations.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Clarissa Feltrin et al.
Chemical research in toxicology, 33(9), 2408-2419 (2020-08-14)
The use of medicinal plants concomitantly with conventional drugs can result in herb-drug interactions that cause fluctuations in drug bioavailability and consequent therapeutic failure and/or toxic effects. The CYP superfamily of enzymes plays an important role in herb-drug interactions. Among

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service