Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

E9256

Sigma-Aldrich

Erythromycin stearate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H67NO13 · C18H36O2
CAS Number:
Molecular Weight:
1018.40
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

biological source

synthetic

form

solid

color

white to off-white

solubility

ethanol: 50 mg/mL, clear to very slightly hazy, colorless

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

mode of action

protein synthesis | interferes

storage temp.

2-8°C

SMILES string

CCCCCCCCCCCCCCCCCC(O)=O.CC[C@@H]1OC(=O)[C@@H](C)[C@H](O[C@H]2C[C@@](C)(OC)[C@@H](O)[C@H](C)O2)[C@@H](C)[C@H](O[C@@H]3O[C@H](C)C[C@@H]([C@H]3O)N(C)C)[C@@](C)(O)C[C@H](C)C(=O)C(C)[C@@H](O)[C@]1(C)O

InChI

1S/C37H67NO13.C18H36O2/c1-14-25-37(10,45)30(41)20(4)27(39)18(2)16-35(8,44)32(51-34-28(40)24(38(11)12)15-19(3)47-34)21(5)29(22(6)33(43)49-25)50-26-17-36(9,46-13)31(42)23(7)48-26;1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h18-26,28-32,34,40-42,44-45H,14-17H2,1-13H3;2-17H2,1H3,(H,19,20)/t18-,19+,20?,21+,22-,23-,24-,25-,26-,28+,29+,30+,31-,32-,34-,35-,36+,37?;/m0./s1

InChI key

YAVZHCFFUATPRK-JGJPHTNUSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: macrolide

Application

Erythromycin stearate is used to study leucocyte motility and transformation and on release in vitro of prostaglandin E2 by stimulated leucocytes[1]. It is also used to study abnormal polymorphonuclear leucocyte migration and chemotaxis[2][3].

Biochem/physiol Actions

Erythromycin inhibits protein synthesis (elongation) at the level of transpeptidation (aminoacyl translocation A-site to P-site) by binding to the 50s subunit of the bacterial 70s rRNA complex.
Erythromycin inhibits protein synthesis (elongation) at the level of transpeptidation (aminoacyl translocation A-site to P-site).

pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 2

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G K Sandor et al.
Journal (Canadian Dental Association), 57(4), 321-325 (1991-04-01)
Mitral valve prolapse syndrome (MVPS) is the name given to the heart valve abnormality described by Barlow over two decades ago. This condition is of particular importance to the dentist as these patients are thought to be at risk of
H Kawai et al.
Kansenshogaku zasshi. The Journal of the Japanese Association for Infectious Diseases, 68(11), 1324-1329 (1994-11-01)
Although erythromycin estolate has been fully assessed for pertussis treatment, the evaluation of erythromycin ethylsuccinate and stearate, the main erythromycin preparations used in Japan and the US, is inadequate. We evaluated these preparations to establish an appropriate treatment for pertussis
M V Martin et al.
British dental journal, 183(4), 135-137 (1997-08-23)
To evaluate shortened courses of antibiotics in the management of dentoalveolar abscesses. Prospective clinical study over a 3-year period. Examinations department of the Liverpool University Dental Hospital. 759 patients, with acute dentoalveolar abscesses associated with swelling, and an elevation of
A S Amin et al.
Journal of pharmaceutical and biomedical analysis, 14(11), 1625-1629 (1996-08-01)
A simple and selective method for the determination of erythromycin and its stearate and succinate esters in their pure forms and in pharmaceutical formulations is described. The procedure is based on the formation of a blue-coloured (lambda max = 633
O Paulsen et al.
Scandinavian journal of infectious diseases, 24(2), 219-225 (1992-01-01)
The efficacy and tolerance of roxithromycin 150 mg b.i.d. were compared with those of erythromycin stearate 500 mg b.i.d. in patients with lower respiratory tract infections. Out of 86 patients recruited for the study, 79 were evaluable for tolerance and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service