Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

E8887

Sigma-Aldrich

Big Endothelin 39 porcine

≥97% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C193H289N49O58S5
CAS Number:
Molecular Weight:
4383.98
MDL number:
UNSPSC Code:
12352200

assay

≥97% (HPLC)

form

solid

UniProt accession no.

storage temp.

−20°C

Gene Information

Amino Acid Sequence

Cys-Ser-Cys-Ser-Ser-Leu-Met-Asp-Lys-Glu-Cys-Val-Tyr-Phe-Cys-His-Leu-Asp-Ile-Ile-Trp-Val-Asn-Thr-Pro-Glu-His-Ile-Val-Pro-Tyr-Gly-Leu-Gly-Ser-Pro-Ser-Arg-Ser [Disulfide Bridges: 1-15; 3-11]

Biochem/physiol Actions

Compared to ET-1, big endothelins show less vasoconstrictor activity in vitro but similar pressor effects in vivo

pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 1 Oral

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Itoh et al.
FEBS letters, 231(2), 440-444 (1988-04-25)
A cDNA encoding a human endothelium-derived vasoconstrictor peptide, endothelin, was isolated from a human placenta cDNA library. The nucleotide sequence of this cDNA clone showed that the primary structure of the human preproendothelin has 212 amino acid residues and is
M Yanagisawa et al.
Nature, 332(6163), 411-415 (1988-03-31)
An endothelium-derived 21-residue vasoconstrictor peptide, endothelin, has been isolated, and shown to be one of the most potent vasoconstrictors known. Cloning and sequencing of preproendothelin complementary DNA shows that mature endothelin is generated through an unusual proteolytic processing, and regional

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service