Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

E8783

Sigma-Aldrich

pFLAG-Myc-CMV-20 Expression Vector

Shuttle vector for transient intracellular expression of dual tagged N-terminal Met-FLAG and C-terminal c-myc fusion proteins

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

tag

FLAG® tagged
c-Myc tagged (C-terminal)

grade

Molecular Biology

form

buffered aqueous solution

shipped in

dry ice

storage temp.

−20°C

General description

The pFLAG-Myc-CMV-20 Expression Vector is a 4.7 kb derivative of pCMV51 used to establish transient expression of intracellular, dual-tagged N-terminal Met-FLAG and C-terminal c-myc fusion proteins in mammalian cells. The vector encodes the FLAG® epitope (Asp-Tyr-Lys-Xaa-Xaa-Asp) and a c-myc epitope (EQKLISEEDL) upstream and downstream of the multiple cloning sites, respectively.

The pFLAG-Myc-CMV-20 Expression Vector is a shuttle vector for E. coli and mammalian cells. Efficiency of replication is optimal when using an SV40 T antigen expressing host.

The pFLAG-CMV-2-BAP Control Plasmid is a 6.1 kb derivative of the pCMV5 transient expression vector for intracellular expression of N-terminal Met-FLAG bacterial alkaline phosphatase fusion protein in mammalian cells.

Vector Maps and Sequences

Application

The pFLAG-Myc-CMV-20 Expression Vector is suitable for transient, cytoplasmic expression of C-terminal c-myc N-terminal Met-FLAG® fusion proteins in mammalian cells.

Components

  • pFLAG-Myc-CMV-20 Expression Vector 20 μg (E5651) is supplied as 0.5 mg/ml in 10 mM Tris-HCl (pH 8.0) with 1 mM EDTA.
  • pFLAG-CMV-2-BAP Control Plasmid 20 μg (P5100) is supplied as 0.5 mg/ml in 10 mM Tris-HCl (pH 8.0) with 1 mM EDTA.

Principle

The promoter-regulatory region of the human cytomegalovirus drives transcription of FLAG® and c-myc fusion constructs.

Legal Information

FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany
pFLAG-CMV is a trademark of Sigma-Aldrich Co. LLC
pFLAG-Myc-CMV is a trademark of Sigma-Aldrich Co. LLC

Storage Class

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fei Song et al.
Journal of neuropathology and experimental neurology, 71(2), 104-115 (2012-01-18)
Neuregulin 1 (NRG1) is a neuron-derived trophic molecule that supports axoglial and neuromuscular development through several alternatively spliced isoforms; its possible role in the pathogenesis and progression of amyotrophic lateral sclerosis (ALS) is not known. We analyzed the relationship of
Kei Tamura et al.
Blood, 115(4), 850-859 (2009-12-08)
The transition of red blood cells (RBCs) from primitive to definitive erythropoiesis is conserved across vertebrates. In anuran amphibians, the larval RBCs from primitive erythropoiesis are replaced by adult RBCs from definitive erythropoiesis during metamorphosis. The molecular mechanisms by which
Ryo Yokomizo et al.
Stem cell research & therapy, 12(1), 130-130 (2021-02-14)
Thin endometrium adversely affects reproductive success rates with fertility treatment. Autologous transplantation of exogenously prepared endometrium can be a promising therapeutic option for thin endometrium; however, endometrial epithelial cells have limited expansion potential, which needs to be overcome in order

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service