Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

E8772

Sigma-Aldrich

pVL1393 Baculovirus transfer vector

lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200

form

lyophilized powder

storage temp.

−20°C

Specificity

The MCS includes restriction sites for BamHI, BglII, EcoRI, NotI, PstI, SmaI, XbaI, and XmaIII. The two vectors, pVL1392 and pVL1393 have their MCS in opposite orientations to simplify subcloning.

Features and Benefits

9.8-kb Supercoiled vectors that contain recombination sequences homologous to the baculovirus genome. This allows the cloned gene to replace the baculovirus wild-type AcNPV polyhedrin gene; thus, protein expression is controlled by the polyhedrin promoter.

Caution

The insert must provide the translational start signal for the non-fusion recombinant protein.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service