E8402
EphA5/Fc Chimera from rat
>95% (SDS-PAGE), recombinant, expressed in NSO cells, lyophilized powder
Synonym(s):
Bsk, Cek7, Ehk1, Hek7, Rek7
About This Item
biological source
rat
recombinant
expressed in NSO cells
assay
>95% (SDS-PAGE)
form
lyophilized powder
mol wt
monomer calculated mol wt 84 kDa
~110 kDa by SDS-PAGE (reducing)
packaging
pkg of 200 μg
storage condition
avoid repeated freeze/thaw cycles (Do not store in a frost-free freezer.)
technique(s)
ligand binding assay: suitable
impurities
endotoxin, tested
UniProt accession no.
storage temp.
−20°C
Gene Information
rat ... Epha5(79208)
General description
Biochem/physiol Actions
Physical form
Analysis Note
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service